Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648390.1 | XP_007646580.1 | 7 - 8 | 1216 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl3| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648390.1 | XP_007646580.1 | 7 - 8 | 1216 | 84 | |