Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648390.1 | XP_007646580.1 | 8 - 9 | 1288 | 77 | |
| XM_007648391.1 | XP_007646581.1 | 7 - 8 | 1203 | 77 | ||
More Information
Set Membership: |
|

Pvrl3
Genomic Map