Assay Details
| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646159.1 | XP_007644349.1 | 2 - 3 | 1003 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Onecut1| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646159.1 | XP_007644349.1 | 2 - 3 | 1003 | 94 | |
Set Membership: |
|