Assay Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644157.1 | XP_007642347.1 | 7 - 8 | 759 | 57 | |
| GenBank mRNA | JI870747.1 | - | 718 | 57 | ||
More Information
Set Membership: |
|

Sec61a1
Genomic Map