Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648386.1 | XP_007646576.1 | 1 - 2 | 88 | 99 | |
| XM_007648390.1 | XP_007646580.1 | 1 - 2 | 88 | 99 | ||
| XM_007648391.1 | XP_007646581.1 | 1 - 2 | 88 | 99 | ||

Pvrl3
Genomic Map