Assay Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494914.2 | XP_003494962.1 | 13 - 14 | 1593 | 67 | |
| GenBank mRNA | JI890854.1 | - | 1 - 2 | 50 | 67 | |
| JP042297.1 | - | 10 - 11 | 1135 | 67 | ||
More Information
Set Membership: |
|

Larp4b
Genomic Map