Assay Details
| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498271.2 | XP_003498319.1 | 5 - 6 | 683 | 67 | |
| XM_007625416.1 | XP_007623606.1 | 4 - 5 | 584 | 67 | ||
| GenBank mRNA | JI878641.1 | - | 5 - 6 | 726 | 67 | |
| JP044412.1 | - | 5 - 6 | 726 | 67 | ||
More Information
Set Membership: |
|

Eif2s1
Genomic Map