Assay Details
| Gene Symbol: Jmjd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501558.2 | XP_003501606.1 | 8 - 9 | 923 | 58 | |
| XM_007642139.1 | XP_007640329.1 | 8 - 9 | 683 | 58 | ||
| GenBank mRNA | JI879331.1 | - | 3 - 4 | 323 | 58 | |
More Information
Set Membership: |
|

Jmjd8
Genomic Map