Assay Details
| Gene Symbol: Septin-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506340.2 | XP_003506388.2 | 6 - 7 | 573 | 97 | |
| GenBank mRNA | JI888338.1 | - | 1 - 2 | 104 | 97 | |
More Information
Set Membership: |
|

Septin-12
Genomic Map