Assay Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509759.2 | XP_003509807.1 | 3 - 4 | 198 | 74 | |
| XM_007650404.1 | XP_007648594.1 | 3 - 4 | 664 | 74 | ||
| XM_007650405.1 | XP_007648595.1 | 2 - 3 | 239 | 74 | ||
| XM_007650406.1 | XP_007648596.1 | 3 - 4 | 195 | 74 | ||
| XM_007650407.1 | XP_007648597.1 | 3 - 4 | 174 | 74 | ||
| XM_007650409.1 | XP_007648599.1 | 2 - 3 | 165 | 74 | ||
| XM_007650410.1 | XP_007648600.1 | 2 - 3 | 160 | 74 | ||
| XM_007650411.1 | XP_007648601.1 | 2 - 3 | 144 | 74 | ||
| XM_007650412.1 | XP_007648602.1 | 2 - 3 | 142 | 74 | ||
| GenBank mRNA | JI881560.1 | - | 3 - 4 | 159 | 74 | |
More Information
Set Membership: |
|

Tmem80
Genomic Map