Assay Details
| Gene Symbol: Mms22l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647696.1 | XP_007645886.1 | 22 - 23 | 3496 | 65 | |
| GenBank mRNA | JI891923.1 | - | 6 - 7 | 891 | 65 | |
More Information
Set Membership: |
|

Mms22l
Genomic Map