Assay Details
| Gene Symbol: LOC100772034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654401.1 | XP_007652591.1 | 9 - 10 | 1422 | 86 | |
| GenBank mRNA | JI882272.1 | - | 2 - 3 | 263 | 86 | |
| JP040158.1 | - | 2 - 3 | 263 | 86 | ||
More Information
Set Membership: |
|

LOC100772034
Genomic Map