Assay Details
| Gene Symbol: LOC100759342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646389.1 | XP_007644579.1 | 3 - 4 | 710 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759342| Gene Symbol: LOC100759342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646389.1 | XP_007644579.1 | 3 - 4 | 710 | 63 | |
Set Membership: |
|