Assay Details
| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651810.1 | XP_007650000.1 | 7 - 8 | 663 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk32b| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651810.1 | XP_007650000.1 | 7 - 8 | 663 | 57 | |
Set Membership: |
|