Assay Details
| Gene Symbol: LOC100771018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646330.1 | XP_007644520.1 | 9 - 10 | 1147 | 100 | |
| XM_007646331.1 | XP_007644521.1 | 9 - 10 | 1175 | 100 | ||
More Information
Set Membership: |
|

LOC100771018
Genomic Map