Assay Details
| Gene Symbol: Nfe2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007629800.1 | XP_007627990.1 | 3 - 4 | 240 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfe2l3| Gene Symbol: Nfe2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007629800.1 | XP_007627990.1 | 3 - 4 | 240 | 69 | |
Set Membership: |
|