Assay Details
| Gene Symbol: Clvs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504213.2 | XP_003504261.1 | 3 - 4 | 671 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clvs2| Gene Symbol: Clvs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504213.2 | XP_003504261.1 | 3 - 4 | 671 | 75 | |
Set Membership: |
|