Assay Details
| Gene Symbol: Ptges | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513976.2 | XP_003514024.1 | 2 - 3 | 378 | 99 | |
| XM_007654197.1 | XP_007652387.1 | 3 - 4 | 607 | 99 | ||
| XM_007654198.1 | XP_007652388.1 | 2 - 3 | 471 | 99 | ||
More Information
Set Membership: |
|

Ptges
Genomic Map