Assay Details
| Gene Symbol: Cmpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007611594.1 | XP_007609784.1 | 1 - 2 | 150 | 65 | |
| GenBank mRNA | JI883314.1 | - | 1 - 2 | 328 | 65 | |
More Information
Set Membership: |
|

Cmpk1
Genomic Map