Assay Details
| Gene Symbol: Pno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642996.1 | XP_007641186.1 | 3 - 4 | 482 | 65 | |
| GenBank mRNA | JP047894.1 | - | 3 - 4 | 485 | 65 | |
More Information
Set Membership: |
|

Pno1
Genomic Map