Assay Details
| Gene Symbol: Rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498437.2 | XP_003498485.1 | 5 - 6 | 1072 | 95 | |
| XM_007627999.1 | XP_007626189.1 | 5 - 6 | 1000 | 95 | ||
| XM_007628007.1 | XP_007626197.1 | 5 - 6 | 993 | 95 | ||
| XM_007628014.1 | XP_007626204.1 | 4 - 5 | 849 | 95 | ||
| GenBank mRNA | JI870444.1 | - | 1 - 2 | 114 | 95 | |
| JP037698.1 | - | 1 - 2 | 113 | 95 | ||
More Information
Set Membership: |
|

Rnf34
Genomic Map