Assay Details
Gene Symbol: Atox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499376.2 | XP_003499424.1 | 2 - 3 | 214 | 80 | |
GenBank mRNA | JI879305.1 | - | 2 - 3 | 209 | 80 | |
JP041124.1 | - | 2 - 3 | 209 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
