Assay Details
| Gene Symbol: Pop5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503531.2 | XP_003503579.1 | 4 - 5 | 425 | 73 | |
| GenBank mRNA | JI893728.1 | - | 3 - 4 | 398 | 73 | |
| JP054066.1 | - | 3 - 4 | 398 | 73 | ||
More Information
Set Membership: |
|

Pop5
Genomic Map