Assay Details
| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506083.2 | XP_003506131.1 | 4 - 5 | 995 | 67 | |
| XM_007646553.1 | XP_007644743.1 | 4 - 5 | 353 | 67 | ||
| GenBank mRNA | JP059673.1 | - | 2 - 3 | 190 | 67 | |
More Information
Set Membership: |
|

Nsrp1
Genomic Map