Assay Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506547.2 | XP_003506595.2 | 6 - 7 | 594 | 76 | |
| GenBank mRNA | JI891152.1 | - | 6 - 7 | 594 | 76 | |
| JP048474.1 | - | 6 - 7 | 594 | 76 | ||
More Information
Set Membership: |
|

Naa10
Genomic Map