Assay Details
| Gene Symbol: Irak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512135.2 | XP_003512183.1 | 9 - 10 | 1338 | 76 | |
| XM_007652621.1 | XP_007650811.1 | 9 - 10 | 1341 | 76 | ||
| XM_007652622.1 | XP_007650812.1 | 9 - 10 | 1755 | 76 | ||
| GenBank mRNA | JI881828.1 | - | 1 - 2 | 180 | 76 | |
| JP052098.1 | - | 1 - 2 | 180 | 76 | ||
More Information
Set Membership: |
|

Irak4
Genomic Map