Assay Details
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515560.2 | XP_003515608.1 | 4 - 5 | 518 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
