Assay Details
| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646917.1 | XP_007645107.1 | 3 - 4 | 302 | 64 | |
| GenBank mRNA | JP050272.1 | - | 3 - 4 | 330 | 64 | |
More Information
Set Membership: |
|

Rras2
Genomic Map