Assay Details
| Gene Symbol: Cers6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495931.2 | XP_003495979.1 | 6 - 7 | 641 | 63 | |
| XM_007645299.1 | XP_007643489.1 | 6 - 7 | 641 | 63 | ||
| XM_007645309.1 | XP_007643499.1 | 6 - 7 | 641 | 63 | ||
| GenBank mRNA | JI888216.1 | - | 6 - 7 | 553 | 63 | |
| JP046251.1 | - | 6 - 7 | 549 | 63 | ||
More Information
Set Membership: |
|

Cers6
Genomic Map