Assay Details
| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504655.2 | XP_003504703.1 | 1 - 2 | 264 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlx| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504655.2 | XP_003504703.1 | 1 - 2 | 264 | 70 | |
Set Membership: |
|