Assay Details
| Gene Symbol: Rcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607529.1 | XP_007605719.1 | 3 - 4 | 641 | 61 | |
| GenBank mRNA | JI869286.1 | - | 3 - 4 | 522 | 61 | |
More Information
Set Membership: |
|

Rcn3
Genomic Map