Assay Details
| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652200.1 | XP_007650390.1 | 4 - 5 | 613 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp12| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652200.1 | XP_007650390.1 | 4 - 5 | 613 | 69 | |
Set Membership: |
|