Assay Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653052.1 | XP_007651242.1 | 4 - 5 | 389 | 66 | |
| GenBank mRNA | JP037758.1 | - | 4 - 5 | 356 | 66 | |
More Information
Set Membership: |
|

Rab22a
Genomic Map