Assay Details
| Gene Symbol: Ly6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654283.1 | XP_007652473.1 | 3 - 4 | 308 | 82 | |
| XM_007654284.1 | XP_007652474.1 | 3 - 4 | 306 | 82 | ||
More Information
Set Membership: |
|

Ly6e
Genomic Map