Assay Details
| Gene Symbol: Ca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505009.2 | XP_003505057.1 | 10 - 11 | 1353 | 68 | |
| GenBank mRNA | JI868912.1 | - | 5 - 6 | 474 | 68 | |
| JP063035.1 | - | 5 - 6 | 474 | 68 | ||
More Information
Set Membership: |
|

Ca9
Genomic Map