Assay Details
| Gene Symbol: LOC100752113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640245.1 | XP_007638435.1 | 11 - 12 | 2152 | 122 | |
| XM_007640246.1 | XP_007638436.1 | 10 - 11 | 2120 | 122 | ||
| XM_007640247.1 | XP_007638437.1 | 11 - 12 | 2065 | 122 | ||
| XM_007640248.1 | XP_007638438.1 | 11 - 12 | 2152 | 122 | ||
More Information
Set Membership: |
|

LOC100752113
Genomic Map