Assay Details
| Gene Symbol: LOC100752113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640245.1 | XP_007638435.1 | 3 - 4 | 247 | 106 | |
| XM_007640246.1 | XP_007638436.1 | 2 - 3 | 215 | 106 | ||
| XM_007640247.1 | XP_007638437.1 | 3 - 4 | 247 | 106 | ||
| XM_007640248.1 | XP_007638438.1 | 3 - 4 | 247 | 106 | ||
| XM_007640249.1 | XP_007638439.1 | 3 - 4 | 247 | 106 | ||
| XM_007640250.1 | XP_007638440.1 | 3 - 4 | 247 | 106 | ||
| XM_007640251.1 | XP_007638441.1 | 3 - 4 | 247 | 106 | ||
| XM_007640252.1 | XP_007638442.1 | 3 - 4 | 247 | 106 | ||
More Information
Set Membership: |
|

LOC100752113
Genomic Map