Assay Details
| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI878183.1 | - | 1 - 2 | 184 | 109 | |
| JP038693.1 | - | 1 - 2 | 184 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763249| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI878183.1 | - | 1 - 2 | 184 | 109 | |
| JP038693.1 | - | 1 - 2 | 184 | 109 | ||
Set Membership: |
|