Assay Details
| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653051.1 | XP_007651241.1 | 5 - 6 | 830 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp5| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653051.1 | XP_007651241.1 | 5 - 6 | 830 | 79 | |
Set Membership: |
|