Assay Details
| Gene Symbol: Zzz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650153.1 | XP_007648343.1 | 6 - 7 | 2156 | 72 | |
| XM_007650158.1 | XP_007648348.1 | 6 - 7 | 2153 | 72 | ||
| GenBank mRNA | JI872323.1 | - | 7 - 8 | 2360 | 72 | |
| JP064449.1 | - | 7 - 8 | 2359 | 72 | ||
More Information
Set Membership: |
|

Zzz3
Genomic Map