Assay Details
| Gene Symbol: LOC100775001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653079.1 | XP_007651269.1 | 4 - 5 | 591 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100775001| Gene Symbol: LOC100775001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653079.1 | XP_007651269.1 | 4 - 5 | 591 | 92 | |