Assay Details
| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653761.1 | XP_007651951.1 | 7 - 8 | 1161 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763249| Gene Symbol: LOC100763249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653761.1 | XP_007651951.1 | 7 - 8 | 1161 | 81 | |
Set Membership: |
|