Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496903.2 | XP_003496951.1 | 7 - 8 | 694 | 67 | |
| GenBank mRNA | JI895510.1 | - | 7 - 8 | 654 | 67 | |
| JP052736.1 | - | 7 - 8 | 654 | 67 | ||
More Information
Set Membership: |
|

Dcxr
Genomic Map