Assay Details
| Gene Symbol: Dhodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501067.2 | XP_003501115.1 | 8 - 9 | 1195 | 59 | |
| XM_007641769.1 | XP_007639959.1 | 8 - 9 | 1172 | 59 | ||
| GenBank mRNA | JI876814.1 | - | 3 - 4 | 390 | 59 | |
| JP057141.1 | - | 3 - 4 | 390 | 59 | ||
More Information
Set Membership: |
|

Dhodh
Genomic Map