Assay Details
| Gene Symbol: Acsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501384.2 | XP_003501432.1 | 7 - 8 | 990 | 53 | |
| XM_007642034.1 | XP_007640224.1 | 7 - 8 | 990 | 53 | ||
| XM_007642035.1 | XP_007640225.1 | 7 - 8 | 991 | 53 | ||
| XM_007642036.1 | XP_007640226.1 | 7 - 8 | 992 | 53 | ||
More Information
Set Membership: |
|

Acsf2
Genomic Map