Assay Details
| Gene Symbol: Nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501491.2 | XP_003501539.1 | 2 - 3 | 211 | 56 | |
| XM_007642188.1 | XP_007640378.1 | 2 - 3 | 196 | 56 | ||
More Information
Set Membership: |
|

Nme3
Genomic Map