Assay Details
| Gene Symbol: Atp6v1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504676.1 | XP_003504724.1 | 1 - 2 | 88 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g3| Gene Symbol: Atp6v1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504676.1 | XP_003504724.1 | 1 - 2 | 88 | 70 | |
Set Membership: |
|