Assay Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508059.2 | XP_003508107.1 | 2 - 3 | 219 | 69 | |
| XM_003508060.2 | XP_003508108.1 | 2 - 3 | 245 | 69 | ||
| XM_007648691.1 | XP_007646881.1 | 2 - 3 | 219 | 69 | ||
| GenBank mRNA | JP049887.1 | - | 2 - 3 | 196 | 69 | |
More Information
Set Membership: |
|

Hus1
Genomic Map