Assay Details
| Gene Symbol: Ces3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508532.1 | XP_003508580.1 | 4 - 5 | 572 | 58 | |
| XM_003508533.1 | XP_003508581.1 | 4 - 5 | 572 | 58 | ||
| XM_003508534.1 | XP_003508582.1 | 4 - 5 | 572 | 58 | ||
More Information
Set Membership: |
|

Ces3
Genomic Map