Assay Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640871.1 | XP_007639061.1 | 1 - 2 | 167 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam173b| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640871.1 | XP_007639061.1 | 1 - 2 | 167 | 73 | |